8 Jul 2026
How to Load 3D Image Data into Amira-Avizo Software
Nearly all Amira-Avizo projects begin by loading image data. This brief tutorial explains how to load 2D and 3D image data from disk.
This tutorial shows typical ways to load image data into Amira and Avizo Software. There are few agreed upon file formats for 3D image data, so the software is flexible but sometimes requires user input. The video covers the "Open Data" and "Open Data As" dialogues as well as Image Read Parameters for loading a series of 2D images as a 3D dataset.