EZ-Link™ Sulfo-NHS-LC-Biotin, No-Weigh™ Format, 10 x 1 mg - FAQs
View additional product information for EZ-Link™ Sulfo-NHS-LC-Biotin - FAQs (21335, A39257)
4 product FAQs found
Can I dissolve the EZ-Link Sulfo-NHS-LC-Biotin, No-Weigh Format (Cat. No. A39257, 21335) in water for storage?
No, the EZ-Link Sulfo-NHS-LC-Biotin, No-Weigh Format (Cat. No. A39257) should not be dissolved in water for storage. We recommended storing EZ-Link Sulfo-NHS-LC-Biotin, No-Weigh Format at -20 degrees C with desiccant as it is very moisture sensitive. Furthermore, to avoid moisture condensation onto the product, equilibrate vial to room temperature before opening.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can you provide the shelf-life for EZ-Link Sulfo-NHS-LC-Biotin?
EZ-Link Sulfo-NHS-LC-Biotin is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (-20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Can you provide the shelf life for EZ-Link Sulfo-NHS-LC-Biotin, No-Weigh Format?
EZ-Link Sulfo-NHS-LC-Biotin, No-Weigh Format is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (desiccated, at -20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
How does a product with "No-Weigh" format reduce my concern over reagent stability?
The No-Weigh product format, where small amounts of reagent are pre-weighed and packaged separately, prevents the contamination and loss of reagent reactivity over time associated with repetitive opening and closing of a single vial. This format enables use of a fresh vial of reagent each time and eliminates the hassle of weighing out small amounts of reagent. Please view additional information and the video at: https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-labeling-crosslinking/no-weigh-packaging.html
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.