Search
Search

ATB BETA| Gene Symbol: ATB BETA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101634.4 | NP_564033.1 | 1 - 2 | 481 | 88 | |
| NM_179350.4 | NP_849681.1 | 1 - 2 | 485 | 88 | ||
| GenBank mRNA | AF370519.1 | - | 1 - 2 | 375 | 88 | |
| AK226540.1 | - | 1 - 2 | 425 | 88 | ||
| AY045665.1 | - | 1 - 2 | 372 | 88 | ||
| BT000645.1 | - | 1 - 2 | 107 | 88 | ||
| BT002183.1 | - | 1 - 2 | 107 | 88 | ||
| U40161.1 | - | 1 - 2 | 193 | 88 | ||
Set Membership: |
|