Search
Search

APK2B| Gene Symbol: APK2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126335.6 | NP_178383.1 | 1 - 2 | 406 | 69 | |
| NM_201674.2 | NP_973403.1 | 1 - 2 | 257 | 69 | ||
| GenBank mRNA | AF428432.1 | - | 1 - 2 | 256 | 69 | |
| AY125559.1 | - | 1 - 2 | 65 | 69 | ||
| D88207.1 | - | 1 - 2 | 205 | 69 | ||
Set Membership: |
|