Search
Search

HEN1| Gene Symbol: HEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203853.2 | NP_001190782.1 | 5 - 6 | 1147 | 110 | |
| NM_118209.3 | NP_567616.1 | 4 - 5 | 1292 | 110 | ||
| GenBank mRNA | AF327068.1 | - | 4 - 5 | 1074 | 110 | |
Set Membership: |
|