Search
Search

SGT1A| Gene Symbol: SGT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036627.2 | NP_001031704.1 | 4 - 5 | 597 | 75 | |
| NM_118488.6 | NP_194088.1 | 5 - 6 | 639 | 75 | ||
| NM_179098.3 | NP_849429.1 | 4 - 5 | 723 | 75 | ||
| GenBank mRNA | AY063978.1 | - | 4 - 5 | 594 | 75 | |
| AY096395.1 | - | 4 - 5 | 457 | 75 | ||
Set Membership: |
|