Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148393.3 | NP_680759.1 | 1 | 1203 | 91 | |
GenBank mRNA | AB189464.1 | - | 1 | 1196 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |