Search
Search

PDH-E1 BETA| Gene Symbol: PDH-E1 BETA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102751.5 | NP_174304.1 | 1 - 2 | 311 | 67 | |
| GenBank mRNA | AF361583.1 | - | 1 - 2 | 199 | 67 | |
| AK221898.1 | - | 1 - 2 | 133 | 67 | ||
| AY093988.1 | - | 1 - 2 | 108 | 67 | ||
Set Membership: |
|