Search
Search

MIPS1| Gene Symbol: MIPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120143.4 | NP_195690.1 | 8 - 9 | 1504 | 65 | |
| GenBank mRNA | AF372954.1 | - | 8 - 9 | 1420 | 65 | |
| AY085931.1 | - | 8 - 9 | 1420 | 65 | ||
Set Membership: |
|