Search
Search

TAG1| Gene Symbol: TAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127503.3 | NP_179535.1 | 5 - 6 | 1010 | 89 | |
| GenBank mRNA | AF051849.1 | - | 5 - 6 | 778 | 89 | |
| AJ131831.1 | - | 5 - 6 | 921 | 89 | ||
| AJ238008.1 | - | 5 - 6 | 829 | 89 | ||
| AY054480.1 | - | 5 - 6 | 852 | 89 | ||
| BT008883.1 | - | 5 - 6 | 691 | 89 | ||
Set Membership: |
|