Search
Search

ABCG22| Gene Symbol: ABCG22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036766.2 | NP_001031843.1 | 1 - 2 | 634 | 91 | |
| NM_001342884.1 | NP_001330485.1 | 1 - 2 | 634 | 91 | ||
| NM_120736.3 | NP_568169.1 | 2 - 3 | 566 | 91 | ||
| NM_180450.2 | NP_850781.2 | 1 - 2 | 621 | 91 | ||
| GenBank mRNA | AK317422.1 | - | 1 - 2 | 637 | 91 | |
| AY059787.1 | - | 2 - 3 | 511 | 91 | ||
| AY114068.1 | - | 1 - 2 | 402 | 91 | ||
Set Membership: |
|