Search
Search

GAT1_2.1| Gene Symbol: GAT1_2.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180612.4 | NP_850943.1 | 1 - 2 | 1009 | 78 | |
| GenBank mRNA | AK175150.1 | - | 1 - 2 | 996 | 78 | |
| AK175288.1 | - | 1 - 2 | 992 | 78 | ||
| AK176685.1 | - | 1 - 2 | 996 | 78 | ||
| BT001098.1 | - | 1 - 2 | 922 | 78 | ||
Set Membership: |
|