Search
Search

ERL2| Gene Symbol: ERL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342930.1 | NP_001331523.1 | 23 - 24 | 2011 | 71 | |
| NM_120800.4 | NP_196335.2 | 25 - 26 | 2566 | 71 | ||
| GenBank mRNA | AY244746.1 | - | 25 - 26 | 2396 | 71 | |
| FJ708772.1 | - | 25 - 26 | 2211 | 71 | ||
Set Membership: |
|