Search
Search

AT5G43560| Gene Symbol: AT5G43560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123722.5 | NP_199169.1 | 6 - 7 | 927 | 69 | |
| NM_180794.5 | NP_851125.1 | 5 - 6 | 1257 | 69 | ||
| GenBank mRNA | AY078956.1 | - | 5 - 6 | 1246 | 69 | |
Set Membership: |
|