Search
Search

AT3G56210| Gene Symbol: AT3G56210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084833.2 | NP_001078302.1 | 2 - 3 | 209 | 88 | |
| NM_001084834.2 | NP_001078303.1 | 2 - 3 | 209 | 88 | ||
| NM_001203174.1 | NP_001190103.1 | 2 - 3 | 260 | 88 | ||
| NM_115479.3 | NP_191180.2 | 2 - 3 | 192 | 88 | ||
| GenBank mRNA | AK316901.2 | - | 2 - 3 | 234 | 88 | |
| BT010820.1 | - | 2 - 3 | 170 | 88 | ||
| BT011306.1 | - | 2 - 3 | 95 | 88 | ||
Set Membership: |
|