Search
Search

GRF11| Gene Symbol: GRF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084180.2 | NP_001077649.1 | 3 - 4 | 612 | 72 | |
| NM_103196.4 | NP_564451.2 | 3 - 4 | 612 | 72 | ||
| GenBank mRNA | BT029457.1 | - | 3 - 4 | 467 | 72 | |
Set Membership: |
|