Search
Search

HEMG2| Gene Symbol: HEMG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203378.1 | NP_001190307.1 | 4 - 5 | 362 | 66 | |
| NM_001343324.1 | NP_001331496.1 | 3 - 4 | 676 | 66 | ||
| NM_001343325.1 | NP_001331495.1 | 4 - 5 | 567 | 66 | ||
| NM_001343326.1 | NP_001331494.1 | 4 - 5 | 378 | 66 | ||
| NM_121426.5 | NP_196926.2 | 4 - 5 | 341 | 66 | ||
| GenBank mRNA | AY075665.1 | - | 4 - 5 | 316 | 66 | |
| AY101523.1 | - | 4 - 5 | 283 | 66 | ||
Set Membership: |
|