Search
Search

AtATG18a| Gene Symbol: AtATG18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035841.4 | NP_001030918.4 | 2 - 3 | 1186 | 64 | |
| NM_116142.6 | NP_567132.4 | 2 - 3 | 1197 | 64 | ||
| GenBank mRNA | AY039545.1 | - | 2 - 3 | 1186 | 64 | |
| AY059169.1 | - | 2 - 3 | 1064 | 64 | ||
Set Membership: |
|