Search
Search

PIPK10| Gene Symbol: PIPK10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340304.1 | NP_001329151.1 | 4 - 5 | 855 | 69 | |
| NM_001340305.1 | NP_001329152.1 | 5 - 6 | 739 | 69 | ||
| NM_116349.1 | NP_192028.1 | 4 - 5 | 583 | 69 | ||
Set Membership: |
|