Search
Search

CPK21| Gene Symbol: CPK21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340500.1 | NP_001319867.1 | 2 - 3 | 977 | 71 | |
| NM_116710.3 | NP_192381.1 | 2 - 3 | 1022 | 71 | ||
| GenBank mRNA | AY150502.1 | - | 2 - 3 | 849 | 71 | |
Set Membership: |
|