Search
Search

HPR| Gene Symbol: HPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198420.1 | NP_001185349.1 | 11 - 12 | 1249 | 57 | |
| NM_105471.4 | NP_176968.1 | 11 - 12 | 1249 | 57 | ||
| GenBank mRNA | AF370221.1 | - | 11 - 12 | 1089 | 57 | |
| AK226564.1 | - | 1066 | 57 | |||
| AY099552.1 | - | 11 - 12 | 1081 | 57 | ||
| AY113871.1 | - | 11 - 12 | 1024 | 57 | ||
| BT001237.1 | - | 11 - 12 | 1024 | 57 | ||
Set Membership: |
|