Search
Search

PATL1| Gene Symbol: PATL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105873.3 | NP_177360.1 | 3 - 4 | 1770 | 67 | |
| GenBank mRNA | AY045913.1 | - | 3 - 4 | 1520 | 67 | |
| AY113865.1 | - | 3 - 4 | 1473 | 67 | ||
Set Membership: |
|