Search
Search

HTH| Gene Symbol: HTH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160997.1 | NP_001154469.1 | 4 - 5 | 1263 | 84 | |
| NM_105955.4 | NP_565050.1 | 4 - 5 | 1281 | 84 | ||
| GenBank mRNA | AB027458.2 | - | 4 - 5 | 1245 | 84 | |
| AY054193.1 | - | 4 - 5 | 1253 | 84 | ||
| BT002648.1 | - | 4 - 5 | 1157 | 84 | ||
Set Membership: |
|