Search
Search

IBR3| Gene Symbol: IBR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111561.5 | NP_187337.2 | 10 - 11 | 1671 | 72 | |
| GenBank mRNA | AY091014.1 | - | 10 - 11 | 1434 | 72 | |
| BT002365.1 | - | 10 - 11 | 1380 | 72 | ||
Set Membership: |
|