Search
Search

ACDO1| Gene Symbol: ACDO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036686.1 | NP_001031763.1 | 1 - 2 | 407 | 64 | |
| NM_119288.3 | NP_194867.2 | 1 - 2 | 439 | 64 | ||
| GenBank mRNA | AY093113.1 | - | 1 - 2 | 406 | 64 | |
| BT008424.1 | - | 1 - 2 | 355 | 64 | ||
Set Membership: |
|