Search
Search

VTC5| Gene Symbol: VTC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124894.5 | NP_200323.1 | 3 - 4 | 780 | 105 | |
| GenBank mRNA | AY063788.1 | - | 3 - 4 | 588 | 105 | |
| AY091285.1 | - | 3 - 4 | 272 | 105 | ||
Set Membership: |
|