Search
Search

UVH6| Gene Symbol: UVH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100201.3 | NP_171818.1 | 8 - 9 | 1937 | 97 | |
| NM_179253.1 | NP_849584.1 | 7 - 8 | 2000 | 97 | ||
| GenBank mRNA | AY062471.1 | - | 8 - 9 | 1937 | 97 | |
| AY090788.1 | - | 7 - 8 | 2000 | 97 | ||
| AY090915.1 | - | 7 - 8 | 1800 | 97 | ||
| BT010344.1 | - | 7 - 8 | 1800 | 97 | ||
Set Membership: |
|