Search
Search

IAA29| Gene Symbol: IAA29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342131.1 | NP_001329439.1 | 1 - 2 | 374 | 66 | |
| NM_119380.3 | NP_567891.1 | 1 - 2 | 405 | 66 | ||
| GenBank mRNA | AB493715.1 | - | 1 - 2 | 258 | 66 | |
| AB493716.1 | - | 1 - 2 | 258 | 66 | ||
| AK229689.1 | - | 1 - 2 | 350 | 66 | ||
| AY669800.1 | - | 1 - 2 | 258 | 66 | ||
Set Membership: |
|