Search
Search

NAI2| Gene Symbol: NAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035631.2 | NP_001030708.1 | 13 - 14 | 869 | 149 | |
| NM_001338191.1 | NP_001326809.1 | 13 - 14 | 869 | 149 | ||
| NM_112465.4 | NP_188216.2 | 15 - 16 | 983 | 149 | ||
| GenBank mRNA | AY120750.1 | - | 15 - 16 | 982 | 149 | |
| BT001207.1 | - | 15 - 16 | 941 | 149 | ||
Set Membership: |
|