Search
Search

AT2G48030| Gene Symbol: AT2G48030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337289.1 | NP_001325107.1 | 1 - 2 | 1019 | 80 | |
| NM_130370.4 | NP_566121.1 | 1 - 2 | 993 | 80 | ||
| GenBank mRNA | AY087825.1 | - | 1 - 2 | 835 | 80 | |
| BT025999.1 | - | 1 - 2 | 694 | 80 | ||
Set Membership: |
|