Search
Search

SEC10| Gene Symbol: SEC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036794.2 | NP_001031871.1 | 5 - 6 | 744 | 77 | |
| NM_001036795.2 | NP_001031872.1 | 5 - 6 | 615 | 77 | ||
| NM_121275.4 | NP_568270.3 | 5 - 6 | 616 | 77 | ||
| GenBank mRNA | AK222187.1 | - | 5 - 6 | 721 | 77 | |
Set Membership: |
|