Search
Search

RCY1| Gene Symbol: RCY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084493.1 | NP_001077962.1 | 4 - 5 | 487 | 120 | |
| NM_001084494.1 | NP_001077963.1 | 3 - 4 | 578 | 120 | ||
| NM_001336070.1 | NP_001324176.1 | 4 - 5 | 494 | 120 | ||
| NM_128200.5 | NP_565622.1 | 4 - 5 | 517 | 120 | ||
| GenBank mRNA | AK226505.1 | - | 4 - 5 | 460 | 120 | |
| AK317155.1 | - | 3 - 4 | 548 | 120 | ||
| AY086218.1 | - | 4 - 5 | 488 | 120 | ||
| AY150419.1 | - | 3 - 4 | 370 | 120 | ||
Set Membership: |
|