Search
Search

HSP60| Gene Symbol: HSP60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113304.5 | NP_189041.1 | 4 - 5 | 857 | 78 | |
| GenBank mRNA | AY099594.1 | - | 4 - 5 | 701 | 78 | |
| BT010398.1 | - | 4 - 5 | 616 | 78 | ||
Set Membership: |
|