Search
Search

CHUP1| Gene Symbol: CHUP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203045.1 | NP_001189974.1 | 8 - 9 | 2948 | 76 | |
| NM_001203046.1 | NP_001189975.1 | 8 - 9 | 2525 | 76 | ||
| NM_001338761.1 | NP_001327702.1 | 9 - 10 | 3133 | 76 | ||
| NM_001338762.1 | NP_001327703.1 | 9 - 10 | 2972 | 76 | ||
| NM_001338763.1 | NP_001327701.1 | 8 - 9 | 3948 | 76 | ||
| NM_113468.5 | NP_189197.2 | 7 - 8 | 3531 | 76 | ||
| GenBank mRNA | AB087408.1 | - | 7 - 8 | 2963 | 76 | |
Set Membership: |
|