Search
Search

KAC2| Gene Symbol: KAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345688.1 | NP_001331380.1 | 2 - 3 | 508 | 69 | |
| NM_001345689.1 | NP_001331383.1 | 2 - 3 | 508 | 69 | ||
| NM_001345690.1 | NP_001331384.1 | 2 - 3 | 508 | 69 | ||
| NM_001345692.1 | NP_001331381.1 | 2 - 3 | 489 | 69 | ||
| NM_125944.3 | NP_201349.3 | 2 - 3 | 318 | 69 | ||
Set Membership: |
|