Search
Search

AT1G12000| Gene Symbol: AT1G12000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101072.4 | NP_172664.1 | 7 - 8 | 956 | 71 | |
| GenBank mRNA | AY062473.1 | - | 7 - 8 | 920 | 71 | |
| AY093260.1 | - | 7 - 8 | 812 | 71 | ||
Set Membership: |
|