Search
Search

ALDH5F1| Gene Symbol: ALDH5F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106592.5 | NP_178062.1 | 1 - 2 | 236 | 81 | |
| GenBank mRNA | AF117335.1 | - | 1 - 2 | 154 | 81 | |
| AF428367.1 | - | 1 - 2 | 156 | 81 | ||
| AY056147.1 | - | 1 - 2 | 142 | 81 | ||
Set Membership: |
|