Search
Search

RIN1| Gene Symbol: RIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122138.3 | NP_197625.1 | 6 - 7 | 1061 | 81 | |
| GenBank mRNA | AY084606.1 | - | 6 - 7 | 975 | 81 | |
| BT000923.1 | - | 6 - 7 | 852 | 81 | ||
Set Membership: |
|