Search
Search

AT1G26330| Gene Symbol: AT1G26330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198170.1 | NP_001185099.1 | 4 - 5 | 480 | 121 | |
| NM_102397.3 | NP_173957.2 | 3 - 4 | 465 | 121 | ||
Set Membership: |
|