Search
Search

VOZ1| Gene Symbol: VOZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084149.1 | NP_001077618.1 | 2 - 3 | 393 | 71 | |
| NM_001332816.1 | NP_001319100.1 | 1 - 2 | 845 | 71 | ||
| NM_001332818.1 | NP_001321335.1 | 2 - 3 | 561 | 71 | ||
| NM_102620.5 | NP_174174.1 | 2 - 3 | 460 | 71 | ||
| GenBank mRNA | AB125256.1 | - | 1 - 2 | 236 | 71 | |
| AK227014.1 | - | 2 - 3 | 352 | 71 | ||
| BT020261.1 | - | 2 - 3 | 311 | 71 | ||
Set Membership: |
|