Search
Search

CDKD1;1| Gene Symbol: CDKD1;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106028.4 | NP_177510.1 | 6 - 7 | 1353 | 95 | |
| GenBank mRNA | AY063843.1 | - | 6 - 7 | 1315 | 95 | |
| AY091227.1 | - | 6 - 7 | 1150 | 95 | ||
Set Membership: |
|