Search
Search

GLK2| Gene Symbol: GLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123786.4 | NP_199232.1 | 4 - 5 | 1171 | 63 | |
| GenBank mRNA | AB062490.1 | - | 4 - 5 | 1017 | 63 | |
| AY026773.1 | - | 4 - 5 | 1017 | 63 | ||
Set Membership: |
|