Search
Search

ATMAK3| Gene Symbol: ATMAK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129369.6 | NP_181348.1 | 5 - 6 | 608 | 80 | |
| NM_201900.2 | NP_973629.1 | 5 - 6 | 551 | 80 | ||
| GenBank mRNA | AK229007.1 | - | 5 - 6 | 503 | 80 | |
| BT010841.1 | - | 4 - 5 | 482 | 80 | ||
| BT012615.1 | - | 3 - 4 | 359 | 80 | ||
Set Membership: |
|