Search
Search

MYB4| Gene Symbol: MYB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120023.3 | NP_195574.1 | 1 - 2 | 764 | 140 | |
| GenBank mRNA | AY070100.1 | - | 1 - 2 | 289 | 140 | |
| AY123004.1 | - | 1 - 2 | 258 | 140 | ||
| AY140037.1 | - | 1 - 2 | 574 | 140 | ||
| AY519615.1 | - | 1 - 2 | 258 | 140 | ||
| BT006302.1 | - | 1 - 2 | 258 | 140 | ||
Set Membership: |
|