Search
Search

BHMT| Gene Symbol: BHMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011679.1 | NP_001011679.1 | 4 - 5 | 530 | 84 | |
| GenBank mRNA | AY854632.1 | - | 4 - 5 | 530 | 84 | |
| BC112599.1 | - | 4 - 5 | 542 | 84 | ||
Set Membership: |
|