Search
Search

SARS2| Gene Symbol: SARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174457.3 | NP_776882.1 | 15 - 16 | 1424 | 80 | |
| GenBank mRNA | AB029947.1 | - | 15 - 16 | 1428 | 80 | |
| BC140548.1 | - | 15 - 16 | 1434 | 80 | ||
| BT021565.1 | - | 15 - 16 | 1423 | 80 | ||
| BT021603.1 | - | 15 - 16 | 1434 | 80 | ||
| BT021873.1 | - | 15 - 16 | 1415 | 80 | ||
| BT021900.1 | - | 15 - 16 | 1429 | 80 | ||
Set Membership: |
|