Search
Search
| Gene Symbol: EMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203361.1 | NP_976237.1 | 4 - 5 | 471 | 85 | |
| GenBank mRNA | AY195839.1 | - | 4 - 5 | 471 | 85 | |
| BC104567.1 | - | 4 - 5 | 528 | 85 | ||
Set Membership: |
|