Search
Search

EXOSC1| Gene Symbol: EXOSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015529.1 | NP_001015529.1 | 7 - 8 | 489 | 90 | |
| GenBank mRNA | BC133440.1 | - | 7 - 8 | 506 | 90 | |
| BT020971.1 | - | 7 - 8 | 489 | 90 | ||
Set Membership: |
|