Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046383.1 | NP_001039848.1 | 4 - 5 | 821 | 70 | |
GenBank mRNA | BC105520.1 | - | 4 - 5 | 821 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |