Search
Search

ARSL| Gene Symbol: ARSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097988.1 | NP_001091457.1 | 8 - 9 | 1284 | 60 | |
| GenBank mRNA | BC140583.1 | - | 8 - 9 | 1284 | 60 | |
| BT030685.1 | - | 8 - 9 | 1390 | 60 | ||
Set Membership: |
|